missing translation for 'onlineSavingsMsg'
Learn More
Carrageenan, iota type Product Code.: 15473349

Carrageenan, iota type

Product Code. 15473349
100 g, 100g
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
This item is not returnable. View return policy

Product Code. 15473349

Brand: Thermo Scientific Alfa Aesar J60603.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Forms soft gels in the presence of calcium ions | CAS: 9062-07-1 | (C12H16O15S2)n | NaN g/mol

Carrageenan, iota type is used as a stabilizer, for suspending cocoa in chocolate manufacture, and to clarify beverages. Carrageenans are used commercially as thickeners and stabilizing agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carrageenan, iota type is used as a stabilizer, for suspending cocoa in chocolate manufacture, and to clarify beverages. Carrageenans are used commercially as thickeners and stabilizing agents.

Solubility
Partially soluble in water, Soluble in hot water. Insoluble in ethanol, in hot concentrated salt solutions.

Notes
Stable under normal conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9062-07-1
Molecular Formula (C12H16O15S2)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00151512
InChI Key QIDSWKFAPCTSKL-RRQHLKGPSA-J
Synonym iota-carrageenans, 1->4-3,6-anhydro-2-o-sulfonato-alpha-d-galactopyranosyl-1->3-4-o-sulfonato-beta-d-galactopyranan
PubChem CID 11966245
IUPAC Name [(2R,3R,4R,5R,6S)-4,5-dihydroxy-6-[[(1R,3R,4R,5S,8S)-3-[(2R,3S,4R,5R,6S)-5-hydroxy-2-(hydroxymethyl)-6-[[(1R,3S,4R,5S,8S)-3-hydroxy-4-sulfonatooxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-3-sulfonatooxyoxan-4-yl]oxy-4-sulfonatooxy-2,6-dioxabicyclo[3.2.1]octShow MoreShow Less
SMILES OC[C@H]1O[C@@H](O[C@H]2[C@H]3CO[C@@H]2[C@@H](OS(O)(=O)=O)[C@@H](-*)O3)[C@H](O)[C@@H](O-*)[C@H]1OS(O)(=O)=O

Specifications

Color Brown to White
Quantity 100 g
Solubility Information Partially soluble in water,Soluble in hot water. Insoluble in ethanol,in hot concentrated salt solutions.
Physical Form Powder
Chemical Name or Material Carrageenan, iota type

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberZ16K008 Certificate TypeCertificate of Analysis Date17/12/2024 Product CodeJ60603.30,J60603.22
Lot NumberQ23K041 Certificate TypeCertificate of Analysis Date11/06/2024 Product CodeJ60603.30,J60603.22
Lot NumberW22J043 Certificate TypeCertificate of Analysis Date06/10/2023 Product CodeJ60603.30,J60603.22
Lot NumberZ12I016 Certificate TypeCertificate of Analysis Date06/01/2023 Product CodeJ60603.30,J60603.22
Lot NumberM24I060 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeJ60603.30,J60603.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Carrageenan, iota type >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.