Learn More
Ratiolab™ Cardboard Cryo Boxes with 6 x 6 Grid
These Cardboard Cryo Boxes with 6 x 6 Grid store samples at ultra-low temperatures with a grid designed to hold 5 mL tubes.
Brand:  Ratiolab™ 5142756
Product Code. 16001492
  • 740.00 NOK / Pack of 10
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Description
  • Temperature-resistant down to -80°C for storing samples at ultra-low temperatures
  • Water-repellent standard coating makes the cardboard highly durable
  • 6 x 6 grid supports 5 mL tubes
Specifications
Specifications
White
Cryo Box
6 x 6 Grid
Non-sterile
136 x 136 x 75 mm
Cardboard
5 mL Tubes
10 Boxes
-80°C