Learn More
Carbonate solution, 0.35M Carb./0.10M Bicarb., for Ion Chromatography, 100mg/L, H2O matrix, Fisher chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Bottle
Specifications
Specifications
Packaging | Bottle |
Solution Contains | 0.35M Carbonate/0.10M Bicarbonate |
Physical Form | Liquid |
Concentration | 100 mg/L |
Type | Single-component Standard Solution |
Solvent or Matrix | H2O |
Quantity | 100 mL |
Chemical Name or Material | Carbonate solution |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1063570 | Certificate of Analysis | 04/03/2025 | A 106 100 |
Product Content Correction
Product Title
Carbonate solution, 0.35M Carb./0.10M Bicarb., for Ion Chromatography, 100mg/L, H2O matrix, Fisher chemical™ >