missing translation for 'onlineSavingsMsg'
Learn More

Carbon tetrabromide, 98% (dry wt.), may cont. up to ca 6% water

CAS: 558-13-4 | CBr4 | 331.627 g/mol

552.00 NOK - 3070.00 NOK

Chemical Identifiers

CAS 558-13-4
Molecular Formula CBr4
Molecular Weight (g/mol) 331.627
MDL Number MFCD00000117
InChI Key HJUGFYREWKUQJT-UHFFFAOYSA-N
Synonym carbon tetrabromide, methane, tetrabromo, carbon bromide, methane tetrabromide, bromid uhlicity, carbontetrabromide, methane, tetrabromide, tetrabrommethan, bromid uhlicity czech
PubChem CID 11205
ChEBI CHEBI:47875
IUPAC Name tetrabromomethane
SMILES C(Br)(Br)(Br)Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11394304
SDS View Documents
Thermo Scientific Alfa Aesar
L00821.14
25 g
552.00 NOK
25g
Please to purchase this item. Need a web account? Register with us today!
11304314
SDS View Documents
Thermo Scientific Alfa Aesar
L00821.22
100 g
955.00 NOK
100g
Please to purchase this item. Need a web account? Register with us today!
11314314
SDS View Documents
Thermo Scientific Alfa Aesar
L00821.36
500 g
3070.00 NOK
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Carbon tetrabromide is widely utilized as a solvent for greases, waxes and oils. In Appel reaction, it is used to convert alcohols to alkyl bromides. It has the property of non-flammability and finds application in fire-resistant chemicals as an active ingredient. It is also used for separating minerals. It can serves as an intermediate in manufacturing agrochemicals. It plays a vital role in the plastic and rubber industries for blowing and vulcanization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbon tetrabromide is widely utilized as a solvent for greases, waxes and oils. In Appel reaction, it is used to convert alcohols to alkyl bromides. It has the property of non-flammability and finds application in fire-resistant chemicals as an active ingredient. It is also used for separating minerals. It can serves as an intermediate in manufacturing agrochemicals. It plays a vital role in the plastic and rubber industries for blowing and vulcanization.

Solubility
Insoluble in water. Soluble in ether, chloroform and ethanol.

Notes
Store in cool place. Incompatible with strong oxidizing agents, strong bases and lithium.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Carbon tetrabromide, 98% (dry wt.), may cont. up to ca 6% water

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.