Learn More
Carbenicillin disodium, MedChemExpress
MedChemExpress Carbenicillin disodium is a beta-lactam penicillin derivative that interference with final stage of bacterial cell wall synthesis.
979.83 NOK - 2939.49 NOK
Specifications
Chemical Name or Material | Carbenicillin disodium |
---|---|
CAS | 4800-94-6 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C17H16N2Na2O6S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16422219
|
Medchem Express
HY-B0525A |
1 g |
979.83 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16440978
|
Medchem Express
HY-B0525A |
5 g |
2939.49 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbenicillin disodium is a beta-lactam penicillin derivative that interference with final stage of bacterial cell wall synthesis.Specifications
Carbenicillin disodium | |
White | |
C17H16N2Na2O6S | |
H2O : 125 mg/mL (295.96 mM; Need ultrasonic) ∣DMSO : 62.5 mg/mL (147.98 mM; Need ultrasonic) | |
422.36 | |
95.0% | |
Solid |
4800-94-6 | |
Research | |
Sodium carbenicillin | |
O=C([C@@H](C(C)(C)S[C@]1([H])[C@@H]2NC(C(C(O[Na])=O)C3=CC=CC=C3)=O)N1C2=O)O[Na] | |
422.36 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
H317∣H334
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture and light∣The compound is unstable in solutions, freshly prepared is recommended.
Recommended Storage : 4°C, sealed storage, away from moisture and light∣The compound is unstable in solutions, freshly prepared is recommended.
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.