missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ FSH Ligand Leakage ELISA

Thermo Scientific™ CaptureSelect™ FSH Ligand Leakage ELISA is designed to detect possible leached ligand of follicle-stimulating hormone (FSH) purified using CaptureSelect FSH Affinity Matrix.

11750.00 NOK - 102895.00 NOK

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Assay ELISA
For Use With (Equipment) Microplate Reader
Product Line CaptureSelect™
Type ELISA Kit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18456508
View Documents
Thermo Scientific™
810318001
1 Assay
11750.00 NOK
Each
Please to purchase this item. Need a web account? Register with us today!
18436718
View Documents
Thermo Scientific™
810318010
10 Assays
102895.00 NOK
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CaptureSelect FSH Ligand Leakage ELISA is designed to detect possible leached ligand of follicle-stimulating hormone (FSH) purified using CaptureSelect FSH Affinity Matrix. The ligand, a 14 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect FSH Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Specifications

Specifications

Store in freezer (-5°C to -30°C).
Microplate Reader
ELISA Kit
ELISA
CaptureSelect™
Kit
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ CaptureSelect™ FSH Ligand Leakage ELISA

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.