Learn More
Marienfeld Superior™ Capillary Tubes for Microhaematocrits
Use Marienfeld Superior™ Capillary tubes for microhaematocrits for the centrifugation of blood samples
323.34 NOK - 336.16 NOK
Specifications
Product Type Capillary Tube
Additive Sodium Heparin
Material Glass
Quantity 10x100
For Use With (Application) Centrifugation of blood samples
View More Specs
Products 2
Product Code Brand Capacity (Metric) Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric) Inner Price Quantity & Availability  
13552140
View Documents
Marienfeld Superior™
2901001
60 ÎĽL 0.9 to 1.0 mm
323.34 NOK
Pack of 1000
13522140
missing translation for 'viewDocuments'
Marienfeld Superior™
2901000
75 ÎĽL 1.1-1.2 mm
336.16 NOK
Pack of 1000
Description
Description
  • Made of high grade quality glass
  • In compliance with DIN ISO 12 772
  • Available with and without heparinization
  • Fire polishing at the color coded end ensures that centrifuges are not damaged
  • For single use
  • 100 pieces in a dispenser vial, 10 vials in a box
  • For in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark, recommended best-before-date and batch number for comprehensive information and traceability
Specifications
Specifications
Capillary Tube
Glass
Centrifugation of blood samples
CE-marked (IVD)
Sodium Heparin
10x100
75 mm
Product Title
Marienfeld Superior™ Capillary Tubes for Microhaematocrits