Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Excellent for melting point analysis in the laboratory. Hirschmann Instruments™ Capillary Tubes for Melting Point Determination, One End Closed are manufactured from clear glass.
Brand:  Hirschmann™ 9212315
 View more versions of this product
Product Code. 10395411
  • 2500.00 NOK / Pack of 1000
Estimated Shipment: 30-06-2025
Description
  • Capillary tubes
  • Available in a range of diameters/lengths
Specifications
Specifications
2.35 mm
One end closed
Capillary Tube
Melting point determination
150 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed > Length: 150mm; O.D.: 2.35mm