Learn More
Calretinin Monoclonal Antibody (DAK Calret 1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512539
Additional Details : Weight : 0.01000kg
Description
MA5-12539 targets Calretinin in IHC (P) applications and shows reactivity with Human samples. The MA5-12539 immunogen is recombinant full length human calretinin protein cloned from Mero-41 cells.
Calretinin is encoded by the CALB2 gene located on chromosome 16. It is a troponin C superfamily protein having six EF-hand structural motifs. The EF motif allows the protein to bind calcium, and function in calcium signaling, calcium buffering, and modulating neuronal excitability. Calretinin is expressed in the mesothelium, mast cells, some neural cells, hair follicles, and fat cells.Specifications
Calretinin | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
P22676, Q08331 | |
CALB2 | |
Recombinant full length human calretinin protein cloned from Mero-41 cells | |
RUO | |
12308, 794 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
DAK Calret 1 | |
Unconjugated | |
CALB2 | |
29 kDa calbindin; 29kD calretinin; 29kDa calbindin; CAB29; CAL2; Calb2; calbindin 2; calbindin 2, (29kD, calretinin); calbindin 2, 29kDa; calbindin D29K; calretinin; CR; testicular secretory protein Li 8 | |
Mouse | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |