Learn More
WTW™ Calcium Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand:  WTW™ 252047
2047.29 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11863921
  • 2145.00 NOK / Each
Specifications
For waste water, drinking water
Certificates
10 to 250 mg/L
Photometric
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers
60
100 Tests
CombiCheck Nitrate (NO3) reagent test kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
WTW™ Calcium Test Kits > Cell Test; Detection Range: 10 to 250mg/L