missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkaline Earth Metal Salts
- Calcium Sulfate Dihydrate, 98.0-102.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Calcium Sulfate Dihydrate, 98.0-102.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals
Product Code. 11450512 Shop All MP Biomedicals Products500 g
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable.
View return policy
Product Code. 11450512
Brand: MP Biomedicals 0219141490
This item is not returnable.
View return policy
Specifications
Chemical Identifiers
CAS | 10101-41-4 |
Molecular Formula | CaH4O6S |
Molecular Weight (g/mol) | 172.16 |
MDL Number | MFCD00149625 |
InChI Key | PASHVRUKOFIRIK-UHFFFAOYSA-L |
Synonym | calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone |
PubChem CID | 24928 |
ChEBI | CHEBI:32583 |
IUPAC Name | calcium dihydrate sulfate |
SMILES | O.O.[Ca++].[O-]S([O-])(=O)=O |
Specifications
Physical Form | Crystalline |
Quantity | 500 g |
Assay Percent Range | 98.0-102.0% |
Formula Weight | 172.2 |
Grade | ACS Reagent |
Chemical Name or Material | Calcium Sulfate Dihydrate |
Product Suggestions
You may be interested in these alternatives
-
Product Code. 10083650
-
Product Code. 11334227
-
Product Code. 11344227
-
Product Code. 11440512
-
Product Code. 11460512
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-5 of 11
-
Catalog No. 15409083
kr 636,00 / Pack of 10 -
Catalog No. 10112640
kr 189,00 / 1kg -
Catalog No. 15429093
kr 610,00 / Pack of 10 -
Catalog No. 10092740
kr 357,30 / 1kg -
Catalog No. 10401841
kr 375,39 / 500mL -
Catalog No. 10776101
kr 590,00 / 1kg -
Catalog No. 11377658
kr 960,00 / 500g -
Catalog No. 10731252
kr 1 210,99 / 500g -
Catalog No. 10161800
kr 307,00 / 500g -
Catalog No. 10158280
kr 372,70 / 500g -
Catalog No. 10021681
kr 623,14 / 500g
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Calcium Sulfate Dihydrate, 98.0-102.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction