Learn More
MP Biomedicals Calcium Free Salt Mixture, Modified
No Calcium in salt mixture.
Brand:  MP Biomedicals 0296020410
 View more versions of this product
Product Code. 11422971
  • 7564.16 NOK / 10kg
Explore more special offers
Description
Description
  • Salt mixture for multiple uses
  • No calcium in mixture
  • Modified to include essential trace minerals
  • This composition is high in phosphate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Not Autoclavable
Single-use
Non-sterile
1 ft.
Animal Diet
10 kg
Room Temperature
0.3048 m
Special Offers
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.