missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNG1 293T Cell Overexpression Lysate (Denatured), Abnova
Brand: Abnova H00000786-T01.100uL
This item is not returnable.
View return policy
Description
Western Blotting
Specifications
293T | |
CACNG1 293T Cell Transient Overexpression Lysate(Denatured) | |
24.53 | |
pCMV-CACNG1 full-length | |
100 μL | |
Human |
Western Blot | |
NM_000727 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-CACNG1 antibody (H00000786-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CACNG1 293T Cell Overexpression Lysate (Denatured), Abnova > 100μL
Spot an opportunity for improvement?Share a Content Correction