missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C7orf26 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21439AF647
This item is not returnable.
View return policy
Description
C7orf26 Polyclonal antibody specifically detects C7orf26 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
C7orf26 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C7orf26 chromosome 7 open reading frame 26 | |
Produced in rabbits immunized with E. coli-derived Human C7orf26 fragment | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
79034 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C7orf26 Antibody, Alexa Fluor™ 647, Novus Biologicals™ > 0.1 mL; Alexa Fluor 647
Spot an opportunity for improvement?Share a Content Correction