Learn More
Uvex™ C500 Cut-Resistant Sleeve
Click to view available options
Size:
Medium
Large
Description
- Bamboo-TwinFlex™ Technology ensures cut protection and abrasion resistance
- Bamboo fiber is soft and silky for increased comfort
- Knit cuff offers a secure fit
- Silicone-free according to the imprint test
- Ideal for applications in the automotive, construction, beverage, glass, and metal industries
Specifications
Specifications
Size | Medium |
Product Type | Cut-resistant Sleeve |
Material | Bamboo Viscose, HPPE, Polyamide |
Color | Green |
Sterility | Non-sterile |
Style | Standard Sleeve |
Disposable | Reusable |
Closure Type | Elastic Ends |
Seam Type | Taped |
Cuff Style | Knit |