missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Burette Stand DUPLEX
Bochem™ Burette Stand DUPLEX is constructed of cast iron and varnished in green.
Brand: Bochem™ 5620
This item is not returnable.
View return policy
Description
- Includes stainless steel rod 18/10 600 x 10mm, 2 plates with diameter 105mm (black and white)
- For buretteholder article no. 5600
Specifications
Cast Iron | |
2 x 105 mm | |
Green Varnished | |
2 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bochem™ Burette Stand DUPLEX > Material: Cast Iron
Spot an opportunity for improvement?Share a Content Correction