missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solutions, BAKER ANALYZED™, J.T. Baker™
Click to view available options
Quantity:
500 mL
1 L
Chemical Name or Material:
Borate
Phosphate
Description
- You can eliminate the time and expense of preparation and standardisation of solutions, helping streamline high-volume analytical work in applications such as quality control
- Contain 0.01% Sodium benzoate
- Certifications: This buffer is traceable to NIST Standard reference buffer
Specifications
Specifications
Certificate of Analysis | Traceable to NIST Standard reference buffer |
pH | 10.00 |
Quantity | 1 L |
Packaging | Plastic bottle |
Chemical Name or Material | Borate |
Safety and Handling

Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362-Take off contaminated clothing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer solutions, BAKER ANALYZED™, J.T. Baker™ >
Spot an opportunity for improvement?Share a Content Correction