missing translation for 'onlineSavingsMsg'
Learn More
Buffer Solution, pH 9.00, NIST, Chem-Lab Product Code.: 11905916

Buffer Solution, pH 9.00, NIST, Chem-Lab

Product Code. 11905916
20 L
Click to view available options
Quantity:
20 L
This item is not returnable. View return policy

Product Code. 11905916

Brand: Chem Lab CL03.0218.9520

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

H3BO3-KCl-NaOH buffer solution for laboratory use and pH measurements, traceable to SRM from NIST

pH = 9.24 at 0°C
pH = 9.11 at 10°C
pH = 9.00 at 20°C
pH = 8.97 at 25°C
pH = 8.93 at 30°C
pH = 8.86 at 40°C
pH = 8.80 at 50°C
pH = 8.75 at 60°C
pH = 8.71 at 70°C
pH = 8.67 at 80°C
pH = 8.64 at 90°C

Specifications

Density 1.01g/mL
pH 9.00
Quantity 20 L
Formulation H3BO3, KCl, NaOH
Packaging EP
Traceability to NIST NIST traceable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution, pH 9.00, NIST, Chem-Lab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.