missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Standard buffer solutions are prepared using gravimetric and volumetric procedures. The batch value is determined by measurement with a combination glass electrode against five-point calibration according to DIN 19268. This pH buffer solution is traceable to Standard Reference Material from NIST.
Specifications
Specifications
Density | 1.01g/cm3 |
pH | 4.00 |
Physical Form | Solution |
Quantity | 250 mL |
Formulation | Per liter: 10.21g Potassium hydrogen phthalate. Contains preservative. |
Traceability to NIST | Traceable to NIST |
Solubility Information | Miscible in water (20°C) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Solution pH 4, Scharlab >
Spot an opportunity for improvement?Share a Content Correction