Learn More
Fisher Chemical Buffer Solution, Colour Coded Red, pH 4, Traceable to NIST, 250ml
Brand: Fisher Chemical J/2826/07
246.81 NOK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Buffer Solution, Colour Coded Red, pH 4, Traceable to NIST, 250ml
Description
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13