Learn More
Buffer concentrate pH 7.00, For 500 ml buffer solution, potassium dihydrogen phosphate / disodium hydrogen phosphate, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Unit Size:
Each
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Chemical Name or Material | Buffer concentrate |
| pH | 7 |
| Quantity | 1 Pc. |
| Packaging | Ampoule |