Learn More
Buffer Colour Coded Concentrated Solution pH7.00 (Phosphate) Yellow, NIST Standard for pH measurement, Fisher Chemical™ Product Code.: 10609643
Buffer Colour Coded Concentrated Solution pH7.00 (Phosphate) Yellow, NIST Standard for pH measurement, Fisher Chemical™
Product Code. 10609643
1 Ampule
Click to view available options
Quantity:
1 Ampule
6 Ea.
Product Code. 10609643
Brand: Fisher Chemical J/2855C/05
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
MDL Number 81857
Specifications
Color Yellow
Physical Form Liquid
Quantity 1 Ampule
Packaging Ampoule
Product Content Correction
Product Title
Buffer Colour Coded Concentrated Solution pH7.00 (Phosphate) Yellow, NIST Standard for pH measurement, Fisher Chemical™ >