Learn More
Bucladesine sodium, MedChemExpress
MedChemExpress Bucladesine sodium salt (Dibutyryl-cAMP sodium salt) is a stabilized cyclic AMP (cAMP) analog and a selective PKA activator. Bucladesine sodium salt raises the intracellular levels of cAMP. Bucladesine sodium salt is also a phosphodiesterase (PDE) inhibitor. Bucladesine sodium salt has anti-inflammatory activity and can be used for impaired wound healing.

821.80 NOK - 1564.57 NOK
Specifications
Chemical Name or Material | Bucladesine sodium |
---|---|
CAS | 16980-89-5 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C18H23N5NaO8P |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16414477
![]() |
Medchem Express
HY-B0764 |
50 mg |
821.80 NOK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16478446
![]() |
Medchem Express
HY-B0764 |
100 mg |
1564.57 NOK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bucladesine sodium salt (Dibutyryl-cAMP sodium salt) is a stabilized cyclic AMP (cAMP) analog and a selective PKA activator. Bucladesine sodium salt raises the intracellular levels of cAMP. Bucladesine sodium salt is also a phosphodiesterase (PDE) inhibitor. Bucladesine sodium salt has anti-inflammatory activity and can be used for impaired wound healing.Specifications
Bucladesine sodium | |
Off-White | |
C18H23N5NaO8P | |
DMSO : 170 mg/mL (345.97 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (101.76 mM) | |
491.37 | |
98.0% | |
Solid |
16980-89-5 | |
Research | |
Dibutyryl cAMP sodium saltDBcAMP sodium salt | |
O=C(CCC)O[C@H]1[C@H](N2C(N=CN=C3NC(CCC)=O)=C3N=C2)O[C@@](CO4)([H])[C@@]1([H])OP4(O[Na])=O | |
491.37 | |
Research |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.