Learn More
Eppendorf™ Bucket for Rotor A-4-38
Bucket designed to hold 16 x 125 mm round bottom tubes.
Brand:  Eppendorf™ 5702764000
5860.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17677635
  • 5883.44 NOK / Each
Description
  • For round bottom tubes
  • Hold 16 x 125 mm tubes
  • For rotor A-4-38
  • Accessory
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotor A-4-38
Adapters, Buckets and Accessories
4/Pk.
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Eppendorf™ Bucket for Rotor A-4-38 > 4/Pk.