Learn More
BST2 Mouse anti-Human, Unconjugated, Clone: 3C4B5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66919-1-IG-150UL
This item is not returnable.
View return policy
Description
The BST2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BST2. This antibody recognizes human antigen. The BST2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
BST2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry | |
3C4B5 | |
Unconjugated | |
150 μL | |
684 | |
IgG2b |