Learn More
BST2 Mouse anti-Human, Unconjugated, Clone: 3C4B5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66919-1-IG-20UL
 View more versions of this product
Product Code. 16849215
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
The BST2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BST2. This antibody recognizes human antigen. The BST2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
BST2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry
3C4B5
Unconjugated
20 μL
684
IgG2b