Learn More
Emma Safety Footwear Bryce XD Safety Shoes
The impenetrable sole protects your feet from sharp and metal objects while the steel toe cap and reinforced PU toe cover offer protection from falling objects. Microfiber construction remains flexible to stand up to cement and resists corrosive substances.
Brand:  Emma Safety Footwear MM33186845
 View more versions of this product
Product Code. 15908371
  • 1605.00 NOK / 1 pair
Estimated Shipment: 12-08-2025
to see stock.
Explore more special offers
Description
  • S3-Safety shoe with sole plate for wet work
  • GripForce™ Duo Heat Resistant Outsole
  • Microfiber construction
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
Bryce XD
Microfiber
45
Product Title
Emma Safety Footwear Bryce XD Safety Shoes > Size: 45