Learn More
Bromothymol Blue, Chem Lab™ Product Code.: 11961367
Estimated Shipment: 04-08-2025
Only null left
Add to basket
Bromothymol Blue, Chem Lab™
Product Code. 11961367
100 mL, 100mL
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 11961367
Brand: Chem Lab CL07.0209.0100
Only null left
Estimated Shipment: 04-08-2025
Add to basket
Only null left
Product Title
Bromothymol Blue, Chem Lab™ >