Learn More
Bromine, 99+%, extra pure Product Code.: 10278312
Bromine, 99+%, extra pure
Product Code. 10278312 Shop All Thermo Scientific Chemicals Products
100 mL, Plastic Coated Glass Bottle, 100mL
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Plastic Coated Glass Bottle
Unit Size:
1 L
100mL
500mL
Product Code. 10278312
Brand: Thermo Scientific Acros 402841000
Request Bulk or Custom Format
CAS: 7726-95-6 | Br | 79.91 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number MFCD00010896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
SMILES [Br-]
Specifications
Color Red to Brown
Physical Form Fuming Liquid
Quantity 100 mL
Assay Percent Range 99+%
Linear Formula Br2
Fieser 03,34; 04,46; 05,55; 06,70; 07,33; 08,52; 09,65; 10,56; 11,75; 12,70; 13,47; 14,56; 15,47
Merck Index 15, 1401
IUPAC Name bromide
Formula Weight 159.82
Percent Purity ≥99.0%
Grade Extra Pure
Packaging Plastic Coated Glass Bottle
Density 3.1110g/mL
Chemical Name or Material Bromine
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0468997 Certificate TypeCertificate of Analysis Date31/01/2025 Product Code402845000,402841000,402840010
Lot NumberA0468737 Certificate TypeCertificate of Analysis Date31/01/2025 Product Code402845000,402841000,402840010
Lot NumberA0461949 Certificate TypeCertificate of Analysis Date11/04/2024 Product Code402845000,402840010,402841000
Lot NumberA0453635 Certificate TypeCertificate of Analysis Date25/08/2023 Product Code402845000,402840010,402841000
Lot NumberA0451155 Certificate TypeCertificate of Analysis Date02/05/2023 Product Code402845000,402840010,402841000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bromine
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Bromine, 99+%, extra pure >