Learn More
Hirschmann™ Bright Lined Counting Chambers
Includes 2 haemocytometer glass covers.
1790.00 NOK - 1985.00 NOK
Specifications
Certifications/Compliance | CE pursuant to IVD 98/79 EC |
---|---|
Includes | 2 haemocytometer glass covers |
Product Code | Brand | Product Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | ||||||
10283861
|
Hirschmann™
8100201 |
Counting Chamber (Burker) |
1790.00 NOK
Each |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
10469132
|
Hirschmann™
8100203 |
Counting Chamber (Neubauer) |
1790.00 NOK
Each |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
10517831
|
Hirschmann™
8100205 |
Counting Chamber (Thoma) |
1790.00 NOK
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. |
||||||
10528961
|
Hirschmann™
8100210 |
Counting Chamber (Fuch-Rosenthal) |
1985.00 NOK
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
||||||
Description
- Double net ruling
- Bright lined
- With LOT number on packaging
- Labelled with: usable until, CE pursuant to IVD 98/79 EC
Specifications
CE pursuant to IVD 98/79 EC | |
2 haemocytometer glass covers |
Product Title
Hirschmann™ Bright Lined Counting Chambers