missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Briggs Chick Salt Mixture No. 5

Salt mixture for chicks. Contains different variety of salts.

1527.46 NOK - 15295.80 NOK

Specifications

Product Type Animal Diet
Autoclavable Not Autoclavable
Disposable Single-use
Sterility Non-sterile
Storage Requirements Room Temperature
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11435632
View Documents
MP Biomedicals
0290283602
2 kg
1527.46 NOK
2kg
Please to purchase this item. Need a web account? Register with us today!
11445632
View Documents
MP Biomedicals
0290283610
10 kg
5751.56 NOK
10kg
Please to purchase this item. Need a web account? Register with us today!
11455632
View Documents
MP Biomedicals
0290283650
50 kg
15295.80 NOK
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Salt mixture containing calcium carbonate, calcium phosphate, etc.
Specifications

Specifications

Animal Diet
Single-use
Room Temperature
0.3048 m
Not Autoclavable
Non-sterile
1 ft.
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.