Learn More
3M™ Breathing Tube, S-222
Click to view available options
:
3M™ Versaflo™ Supplied Air System S-200+
Description
- Spare or replacement part
- Designed for use with 3M™ Versaflo™ Supplied Air System S-200+
- CE approved
- Protective breathing tube covers available to lengthen service life of breathing tubes
Specifications
Specifications
Color | Black |
For Use With (Equipment) | 3 M™ Versaflo™ Supplied Air System S-200+ |
Product Content Correction
Product Title
3M™ Breathing Tube, S-222 >