missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD8 Mouse anti-Human, Clone: 5B6D6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60121-1-IG-20UL
This item is not returnable.
View return policy
Description
Transcriptional repressor that contribute to growth arrest during terminal macrophage differentiation by repressing target genes involved in Ras-dependent proliferation. Represses MMP1 promoter activity.Specifications
BRD8 | |
Monoclonal | |
1.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H0E9 | |
BRD8 | |
BRD8 Fusion Protein Ag0790 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
5B6D6 | |
Unconjugated | |
BRD8 | |
BRD8, bromodomain containing 8, p120, SMAP, SMAP2, TrCP120 | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
10902 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction