Learn More
Buerkle™ Brass Tap
Barrel tap for petrochemical liquids, closed independently when not used (increased safety), thread connection 3/4in.on the outside.
Brand:  Buerkle™ 0535-0010
Product Code. 10011524
  • 570.96 NOK / Each
Only null left
Estimated Shipment: 23-07-2025
Description
  • Cost-effective tap
  • PE, brown
  • Seal made of EPT
Specifications
Specifications
Brass
1 Ea.
Tap
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Buerkle™ Brass Tap > Width: 10mm