Learn More
Witeg™ Brass Cork Borer Sets
Use the Witeg™ Brass Cork Borer Set for your tooling needs.
Brand:  Witeg™ 7 414 305
 View more versions of this product
Product Code. 11897770
  • 11590.00 NOK / Each
Only null left
Estimated Shipment: 22-08-2025
Description
  • Cork
  • Brass
Specifications
Specifications
Bork Borer Set
5 to 26.25 mm
Brass
18Pack
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Witeg™ Brass Cork Borer Sets > Pusher Diameter: 5 to 26.25mm