Learn More
R&L Enterprises™ Cork Borer
Efficiently and cleanly bores holes in cork. R&L Enterprises™ Cork Borers have a cutting tube mounted with safety under-flange in shaped handle.
1750.00 NOK - 2810.00 NOK
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
12863952
|
R&L Enterprises™
3401 |
Range: 4 to 18mm |
1750.00 NOK
Each |
Estimated Shipment: 02-09-2025
Log in to see stock. |
||||||
12873952
|
R&L Enterprises™
3402 |
Range: 4 to 25mm |
2810.00 NOK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
Description
Efficiently and cleanly bores holes in cork. R&L Enterprises™ Cork Borers have a cutting tube mounted with safety under-flange in shaped handle.
- With rod for clearning bores
- Manufactured from brass
- Sizes numbered consecutively from 1 upwards in each set
Specifications
Cork Borer | |
Brass |