Learn More
VACUUBRAND™ Service Kits for NT Series Chemistry Pumps and Systems
Product Code. p-4389395
Click to view available options
For Use With:
MD 4C NT VARIO, PC3003/4 VARIO
ME 4C NT
ME 8C NT
MZ 2C NT/VARIO, PC201 NT, PC3002 VARIO
Unit Size:
Each
Product Code. 16254312
Brand: VACUUBRAND™ 20696864
Only null left
Add to basket
Add to basket
Service kits for VACUUBRAND NT series Chemistry design pumps.
  • Sets include appropriate diaphragms, valves and sealing rings (if applicable)
  • Wrench not included.
TRUSTED_SUSTAINABILITY
Specifications
For Use With ME 4C NT
Type Service Kit