Learn More
BRAND™ HandyStep™ Electronic Repeating Pipetter Accessory, Battery Pack
For use with HandyStep™ Electronic Repeating Pipetter. The HandyStep™ Electronic Repeating Pipetter Accessory, Battery Pack can be easily interchanged for continuous pipetting.
Brand: BRAND™ 705025
512.05 NOK valid until 2025-07-04
Use promo code "24147" to get your promotional price.
Description
- Replaceable nickel-metal-hydride battery pack charges in 2.5 hours (3.5 hours inside instrument)
- Battery pack can be easily interchanged for continuous pipetting
Specifications
NiMH | |
Battery Pack | |
HandyStep™ electronic repeating pipette battery |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59% -
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13