Learn More
BRAND™ Valve Set for seripettor™ Bottle Top Dispensers
Includes 1 filling valve, 1 discharge valve and 2 seals.
Brand:  BRAND™ 6790
260.30 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10103792
  • 389.00 NOK / Each
The Ultimate Lab Appliances Promotion
Use promo code 24075 at checkout
View Available Promotions
Only null left
Estimated Shipment: 10-07-2025
Description
Valve set for seripettor™
  • 1 filling valve
  • 1 discharge valve
  • 2 seals
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Valve Set
Seripettor™ bottletop dispenser
Filling valve, discharge valve and two seals
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.
    Promo Code: 24075
    Expires2025-06-30
    Days
    17
    Hours
    13
    Minutes
    19
    Seconds
    3
Product Content Correction
Product Title
BRAND™ Valve Set for seripettor™ Bottle Top Dispensers > Valve set