Learn More
BRAND™ BRANDplates™ Inlet Opening System PC Membrane Single Inserts
Sterile, individual inserts
Brand: BRAND™ 782867
This item is not returnable.
View return policy
Description
Specifications
Inlet channels | |
24-/6-well BRANDplates™ | |
48 |
PC Membrane 8.0 ÎĽm | |
13 mm |
Product Content Correction
Product Title
BRAND™ BRANDplates™ Inlet Opening System PC Membrane Single Inserts > Type: PC Membrane 8.0 μm