accessibility menu, dialog, popup
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Facility Safety and Maintenance
Signs and Tags
Physical Hazard Signs
Brady™ Bradycone™ Traffic Safety Cone
Print
Brady™ Bradycone™ Traffic Safety Cone
1015.00 NOK - 1240.00 NOK
Specifications
Quantity
1 Ea.
Product Type
Traffic Safety Cone
View More Specs
Products
2
Documents
Product Certifications
Products
2
Product Code
Brand
Height (Metric)
Price
Quantity & Availability
Product Code
Brand
Height (Metric)
Price
Quantity & Availability
11726665
View Documents
missing translation for 'viewProductCertificates'
Brady™
256317
630 mm
1015.00 NOK
Each
Estimated Shipment: 12-12-2024
Log in
to see stock.
Add to basket
11716665
View Documents
missing translation for 'viewProductCertificates'
Brady™
256316
890 mm
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Specifications
Quantity
1 Ea.
Product Type
Traffic Safety Cone
Documents
Product Certifications
Back to Top