Learn More
Brady™ Indoor/Outdoor Permanent Adhesive Vinyl Label Cartridges
Indoor/Outdoor Permanent Adhesive Vinyl Label Cartridges adhere to irregular, rough and curved surfaces.
354.00 NOK - 544.00 NOK
Specifications
Product Type | Label Cartridge |
---|---|
For Use With (Equipment) | Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-LAB Label Printer, BMP21-PLUS Label Printer |
Material | Vinyl |
Print Technology | Thermal Transfer |
Length (English) | 21 ft. |
Product Code | Brand | Color | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Dimensions (L x W) | Price | Quantity & Availability | |||||
11733464
|
Brady™
142794 |
White on Blue | 21 x 0.75 in. (6.4 m x 19.05 mm) |
516.00 NOK
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
12897278
|
Brady™
142797 |
Black on White | 21 x 0.75 in. (6.4 m x 19.05 mm) |
544.00 NOK
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
|||||
15757218
|
Brady™
139744 |
Black on White | 21 x 0.25 in. (6.4 m x 6.35 mm) |
366.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
15359662
|
Brady™
139732 |
White on Purple | 21 x 0.375 in. (6.4 m x 9.53 mm) |
394.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
30041435
|
Brady™
139745 |
Black on Yellow | 21 x 0.25 in. (6.4 m x 6.35 mm) |
354.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
15309672
|
Brady™
139734 |
White on Purple | 21 x 0.75 in. (6.4 m x 19.05 mm) |
516.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11723444
|
Brady™
142800 |
Black on White | 21 x 0.375 in. (6.4 m x 9.53 mm) |
383.00 NOK
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
12802659
|
Brady™
142807 |
Black on White | 21 x 0.5 in. (6.4 m x 12.7 mm) |
442.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11763444
|
Brady™
142802 |
White on Green | 21 x 0.375 in. (6.4 m x 9.53 mm) |
394.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11753444
|
Brady™
142806 |
White on Blue | 21 x 0.375 in. (6.4 m x 9.53 mm) |
394.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11723464
|
Brady™
142799 |
Black on Yellow | 21 x 0.5 in. (6.4 m x 12.7 mm) |
442.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11773444
|
Brady™
142810 |
White on Red | 21 x 0.375 in. (6.4 m x 9.53 mm) |
394.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
12812669
|
Brady™
142809 |
Black on Orange | 21 x 0.375 in. (6.4 m x 9.53 mm) |
394.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
12822659
|
Brady™
142803 |
Black on Yellow | 21 x 0.375 in. (6.4 m x 9.53 mm) |
394.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
12822669
|
Brady™
142796 |
Black on Orange | 21 x 0.5 in. (6.4 m x 12.7 mm) |
442.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
12862659
|
Brady™
142798 |
White on Blue | 21 x 0.5 in. (6.4 m x 12.7 mm) |
442.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
12852669
|
Brady™
142795 |
White on Red | 21 x 0.5 in. (6.4 m x 12.7 mm) |
442.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11743464
|
Brady™
142808 |
White on Green | 21 x 0.75 in. (6.4 m x 19.05 mm) |
516.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11733484
|
Brady™
142804 |
Black on Orange | 21 x 0.75 in. (6.4 m x 19.05 mm) |
516.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11733494
|
Brady™
142811 |
Black on Yellow | 21 x 0.75 in. (6.4 m x 19.05 mm) |
516.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11743484
|
Brady™
142801 |
White on Red | 21 x 0.75 in. (6.4 m x 19.05 mm) |
516.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
Description
Adheres to irregular, rough and curved surfaces
- For use with BMP™21, BMP21-LAB, BMP21-PLUS, IDPal™, LabPal™ Printers (M210, M210-LAB, M211, BMP21-PLUS, BMP21-LAB, IDPAL, LABPAL).
- Made of 8-10 year indoor/outdoor material.
- Matte finish with permanent adhesive.
- Stands up to grease, oil and most industrial chemicals.
- Available in various sizes and colors.
Applications:
General identification, laboratory identification, panel identification, panel labels, plant labels, safety labels, Lean/5S and visual workplace labels, pipe marking, tool labeling, equipment labeling and outdoor labeling.
†Compatible with the BMP™21-PLUS printer only.
Compliance
RoHS Compliant, WEEE, EU REACH SVHC
Specifications
Label Cartridge | |
Vinyl | |
21 ft. | |
0.004 in. | |
Permanent Acrylic | |
Continuous | |
-40°C to +82°C |
Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-LAB Label Printer, BMP21-PLUS Label Printer | |
Thermal Transfer | |
6.4 m | |
0.102 mm | |
Gloss | |
-40°F to +179.6°F |