missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BPIFA2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Specifications
Antigen | BPIFA2 |
---|---|
Concentration | 367 μg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
The BPIFA2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BPIFA2. This antibody recognizes human antigen. The BPIFA2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
BPIFA2 | |
Western Blot, ELISA | |
Unconjugated | |
Human | |
IgG |
367 μg/mL | |
Polyclonal | |
Rabbit | |
140683 | |
Primary |