Learn More
Trajan Scientific and Medical™ BP624 GC Capillary Columns
Used for EPA volatile organics analysis. Trajan Scientific and Medical BP624 GC Capillary Columns are polar phase designed columns.
Brand: Trajan Scientific and Medical™ 054838
This item is not returnable.
View return policy
Description
- Polar phase
- EPA organics analysis (EPA 624, 502.2, SW-846, 8240/8260)
- Ideal for environmental analyses, volatile organics, alcohol analysis
Specifications
0.53 mm | |
Chlorinated Hydrocarbons Analysis, Drinking Water Volatiles, Chlorinated Hydrocarbons, Solvents Analysis | |
3 μm | |
Cyanopropylphenyl Polysiloxane |
BP624 GC Capillary Column | |
0°C to 230/240°C | |
60 m | |
BP624 |