Learn More
Gilac™ Bowls with Handles, 50 Cycle
Click to view available options
Quantity:
10 Pk.
Unit Size:
Each
Description
- Reusable plastic bowl.
- Outstanding durability even in the intensive use cycle.
- Ideal for institutional catering use.
- Pack of 10.
Specifications
Specifications
| Product Type | Bowl |
| Material | Polypropylene |
| Color | White |
| Diameter (Metric) | 134 mm |
| Height (Metric) | 65 mm |
| Quantity | 10 Pk. |
| Weight (Metric) | 0.65 kg |
Product Content Correction
Product Title
Gilac™ Bowls with Handles, 50 Cycle >