man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Specialty Lab Equipment, Instruments, and Apparatuses
Mills
RETSCH Bottom Cones
11924202
Print
RETSCH Bottom Cones
Opening Width: 159mm; For Use With PT 200
Brand:
RETSCH 42.787.0010
View more versions of this product
Product Code.
11924202
30650.00 NOK
/ Each
Quantity
Estimated Shipment: 20-12-2024
Log in
to see stock.
Explore more special offers
Add to basket
This item is not returnable.
View return policy
Specifications
Product Type
Bottom Cone
Diameter (Metric) Opening
159 mm
For Use With (Equipment)
For PT 200