man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Specialty Lab Equipment, Instruments, and Apparatuses
Mills
RETSCH Bottom Cones
10369461
Print
RETSCH Bottom Cones
Opening Width: 110mm; For Use With PK 1000
Brand:
RETSCH 42.787.0003
View more versions of this product
Product Code.
10369461
51465.00 NOK
/ Each
Quantity
Estimated Shipment: 29-01-2025
Log in
to see stock.
Explore more special offers
Add to basket
This item is not returnable.
View return policy
Specifications
Product Type
Bottom Cone
Diameter (Metric) Opening
110 mm
For Use With (Equipment)
For PK 1000