Learn More
Socorex™ Bottle Top Dispenser 530 Model
Brand:  Socorex™ 530.100
6199.08 NOK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
 View more versions of this product
Product Code. 13598290
  • 8195.00 NOK / Each
The Ultimate Lab Appliances Promotion
Use promo code 24075 at checkout
View Available Promotions
Only null left
Estimated Shipment: 22-07-2025
Specifications
Bottletop Dispenser
EN ISO 8655
530
Autoclavable
10 to 100 mL
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.
    Promo Code: 24075
    Expires2025-06-30
Product Content Correction
Product Title
Socorex™ Bottle Top Dispenser 530 Model > Volume Range: 10 to 100mL