Learn More
Gilson™ Bottle Rack
Code rack for bottles
11265.00 NOK - 14170.00 NOK
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
16646135
|
Gilson™
17041950 |
500 mL |
14170.00 NOK
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
16656135
|
Gilson™
17041951 |
1000 mL |
11265.00 NOK
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
Description
- Material: aluminum
- For Kimax or Pyrex bottles
Specifications
Bottle Rack |
Product Title
Gilson™ Bottle Rack