missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Harvey™ Bottle Carrier, 2-Place

Thermo Scientific Harvey™ Bottle Carrier, 2-Place is constructed of polyethylene-coated mild steel and can carry 2 x 2.5L bottles.

Brand:  Thermo Scientific Harvey™ BTS-870-E

Product Code. 12378258

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Polyethylene-coated mild steel
  • Carries 2 x 2.5L bottles of either Wichester pattern or Eurobottle type
  • Maximum diameter of bottle held is 150mm
Specifications

Specifications

Bottle Carrier
Mild Steel
Two 2.5L Winchester or Eurobottle bottles
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers